John Guest Acetal Push-To-Connect Soft Tube Supports
Supplier: VWR International
MFLX34007-39 MFLX34007-40 MFLX34007-38
MFLX34007-39EA 60.04 CAD
MFLX34007-39 MFLX34007-40 MFLX34007-38
John Guest Acetal Push-To-Connect Soft Tube Supports
Fittings and Connectors
No need to buy two fitting halves.
  • Meet NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (More Product Information