John Guest polypropylene threaded elbows
Supplier: VWR International
PP480821W-US PP480822W PP481222W PP481223W
MFLX34007-62EA 55.93 CAD
MFLX34007-62 MFLX34007-63 MFLX34007-64 MFLX34007-65
John Guest polypropylene threaded elbows
Fittings and Connectors
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications; and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.