Stainless Steel Male Threaded to Push-to-Connect Adapters
Supplier: WILLIAM P NUGENT COMPANY INC
PC68SS-42-316 PC68SS-44-316 PC68SS-64-316 MFLX31370-03 MFLX31370-04 MFLX31370-05
MFLX31370-00EA 45.5 CAD
MFLX31370-00 MFLX31370-01 MFLX31370-02 MFLX31370-03 MFLX31370-04 MFLX31370-05
Stainless Steel Male Threaded to Push-to-Connect Adapters
Fittings and Connectors
The ease of push-to-connect and the durability of 316 stainless steel.
  • Constructed of durable 316 stainless steel
The durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines. Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines. Matching material of construction to your stainless piping will reduce the effects of thermal expansion differentials and minimize line leakage.