John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Supplier: VWR International
PI050821S PI050822S PI051222S PI051223S PI051623S PI051624S PM050822S PM050823S PM050421S PM050422S
MFLX34006-66EA 21.65 CAD
MFLX34006-66 MFLX34006-67 MFLX34006-68 MFLX34006-69 MFLX34006-70 MFLX34006-71 MFLX34007-30 MFLX34007-31 MFLX34006-86 MFLX34007-27 MFLX34007-28 MFLX34007-29
John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Fittings and Connectors
No need to buy two fitting halves.
  • Meets NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
  • A single fitting offers secure connections
  • and quick disconnection
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.