Saint Gobain® Plastic Luer-to-Sanitary Adapters
Supplier: VWR International
MFLX31200-50 MFLX31201-50 MFLX40619-96
MFLX31200-50CS 0 CAD
MFLX31200-50 MFLX31201-50 MFLX40619-96
Saint Gobain® Plastic Luer-to-Sanitary Adapters
Fittings and Connectors
Attach with just a twist!
  • Choose between polypropylene (PP) and PVDF material options
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.