Parker High-Pressure Multimedia Solenoid Valves
Supplier: PARKER HANNIFIN PRECISION FLUIDICS
009-0172-900 009-0270-900 009-0269-900 009-0089-900
MFLX01870-01EA 1325.62 CAD
MFLX01870-01 MFLX01870-06 MFLX01870-07 MFLX01870-09 MFLX01870-00 MFLX01870-02 MFLX01870-04 MFLX01870-10 MFLX01870-05 MFLX01870-03
Parker High-Pressure Multimedia Solenoid Valves
Valves Solenoid Valves
Offer outstanding potential for precision control in liquid and gas analysis.
  • High speed — typical response time is <5 ms
  • Maximum pressure up to 1250 psi
  • Constructed of noncorroding, passivated stainless steel
  • Leak tight to 1 x 10-7 cc/sec/atm Helium
These rugged valves operate with high repeatability, combining high speed and high flow in a small size. Coils are rated continuous duty and are potted for protection from dust and dirt.