Air/N2 Gas Mass Flowmeters with Metal Bodies
Supplier: AALBORG INSTRUMENTS & CONTROL
GFMS-010001 GFMS-010003 GFMS-010005 GFMS-010006 GFMS-010008 GFMS-010009 GFMS-011159 GFMS-010013 GFMS-010014 GFMS-010015 GFMS-010018 GFMS-010027 GFMS-010803 MFLX32648-02 MFLX32648-06 MFLX32648-12 MFLX32648-43 MFLX32648-44 MFLX32648-50 MFLX32648-54 MFLX32648-56 MFLX32648-58 MFLX32648-60 MFLX32648-62 MFLX32648-64 MFLX32648-68 MFLX32648-84 MFLX32648-86 MFLX32648-92 MFLX32659-10 MFLX32659-20
MFLX32648-00EA 1145.5 CAD
MFLX32648-00 MFLX32648-04 MFLX32648-08 MFLX32648-10 MFLX32648-14 MFLX32648-16 MFLX32648-19 MFLX32648-34 MFLX32648-36 MFLX32648-42 MFLX32648-45 MFLX32648-66 MFLX32659-00 MFLX32648-02 MFLX32648-06 MFLX32648-12 MFLX32648-43 MFLX32648-44 MFLX32648-50 MFLX32648-54 MFLX32648-56 MFLX32648-58 MFLX32648-60 MFLX32648-62 MFLX32648-64 MFLX32648-68 MFLX32648-84 MFLX32648-86 MFLX32648-92 MFLX32659-10 MFLX32659-20
Air/N2 Gas Mass Flowmeters with Metal Bodies
Flow Meters
Precisely measure flow rates and totals.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 2More Product Information