Grooving Tool for PFA Compression Fittings
Supplier: ANDON SPECIALTIES INC.
GT6M
MFLX31321-49EA 660.44 CAD
MFLX31321-49
Grooving Tool for PFA Compression Fittings
Puller and Inserter Tools
Get a secure seal at higher pressures.
  • Optimize performance of your PFA Compression Fittings
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).