CPC® Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body
Supplier: VWR International
MFLX31413-15 MFLX31309-35 MCD181032 (10PK) LC15004 MC1502 MC181032
MFLX31309-33EA 448.72 CAD
MFLX31309-33 MFLX31309-34 MFLX31309-36 MFLX31413-14 MFLX31413-15 MFLX31309-35 MFLX31413-15BLK MFLX31309-35BLK MFLX31309-34BLK MFLX31309-36BLK
CPC® Metal Quick-Disconnect Fittings, Panel-Mount Threaded Body
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).