CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Supplier: Colder Products
MFLX06353-11 MFLX06353-17 MFLX06353-19 MFLX06353-22 MFLX06353-23 MFLX06353-24 MFLX06353-25 MFLX06353-26 MFLX06353-28 MFLX06353-33 APCD24004BSPT APC17004SH 25PK APC17006SH 25PK APC16006 25PK APCD23006 25PK APCD10004 25PK
MFLX06353-01EA 312.84 CAD
MFLX06353-01 MFLX06353-02 MFLX06353-07 MFLX06353-08 MFLX06353-09 MFLX06353-15 MFLX06353-21 MFLX06353-34 MFLX49007-36 MFLX06353-11 MFLX06353-17 MFLX06353-19 MFLX06353-22 MFLX06353-23 MFLX06353-24 MFLX06353-25 MFLX06353-26 MFLX06353-28 MFLX06353-33 MFLX06353-25BLK MFLX06353-07BLK MFLX06353-08BLK MFLX06353-21BLK MFLX06353-34BLK MFLX06353-01BLK
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Fittings connect in a snap.
  • No exposed metal parts
  • Protects against accidental disconnects
These CPC® APC-Series quick-disconnect fittings, combine a compact design with durable acetal construction, making them an excellent choice for many applications. All fittings feature a 1/4” flow, plastic thumb latch, fewer moving parts, and a smooth contoured design to deliver ease-of-use and excellent flow in a compact size. Simply press the thumb latch to disconnect fitting halves. Choose from in-line compression and hose barb bodies which feature shrouded latches to provide further protection against accidental disconnect. The ferruleless PTF compression fittings do not require ferrules to achieve a secure connection, which makes them easier to use and reuse. PTF fittings are designed for semi-rigid tubing, i.e., polyethylene, nylon, polyurethane, etc.
A complete coupler requires a fitting body and an insert (order separately).