Masterflex® Tuf-Flex® Sanitary Tri-Clamp® Gaskets, Avantor®
Supplier: VWR International
MFLX00278-GL MFLX30547-50 MFLX30547-52 MFLX30547-54 MFLX30547-56 MFLX30547-60 MFLX30547-58
MFLX00278-GLEA 1033.32 CAD
MFLX00278-GL MFLX30547-50 MFLX30547-52 MFLX30547-54 MFLX30547-56 MFLX30547-60 MFLX30547-58
Masterflex® Tuf-Flex® Sanitary Tri-Clamp® Gaskets, Avantor®
Seals Gaskets
Ensure a better process seal with outstanding chemical resistance.
  • The chemical resistance of PTFE and the flexibility of an elastomer
  • Designed for CIP/SIP conditions
The Tuf-Flex gasket grafts a PTFE envelope to an elastomer core. Unlike pure PTFE gaskets, or even other sheathed gaskets, Tuf-Flex will not creep or cold flow over time and therefore maintains the integrity of its seal in your process line. The core gives the gasket the mechanical properties of an elastomer, including memory and flexibility. Designed to meet the stringent requirementsMore Product Information