Guzzler® Portable Diaphragm Hand Pumps
Supplier: BOSWORTH CO
G450-H G400-H COLEP G500-H COLEP G2600
MFLX07090-09EA 290.72 CAD
MFLX07090-09 MFLX07090-10 MFLX07090-11 MFLX07090-12
Guzzler® Portable Diaphragm Hand Pumps
Pumps Drum/Transfer Pumps
Low-cost, high-flow pumping solution.
  • Ideal for pumping shear-sensitive or particulate-laden fluids
  • Durable, portable pumps mount in any position
  • Available in cast aluminum or nonclogging Delrin® acetal plastic
  • 07090-09 has inlet and outlet on same side
Guzzler® hand pumps are designed for quick, easy fluid transfer.
Order service kits, noncollapsing wire-reinforced vinyl hoses, stainless steel hose clamps, and check valves separately in table.