Hedland Spring-Loaded In-Line Flowmeters
Supplier: BADGER METER INC
H625-607 H625-610 H625-616 MFLX03231-01 MFLX03231-03 MFLX03231-07 MFLX03231-21 MFLX03231-33 MFLX03231-41 MFLX03231-43 MFLX03231-45 MFLX03231-47 MFLX03231-51 MFLX03231-61 MFLX03231-67 MFLX03231-09
MFLX03231-23EA 297.04 CAD
MFLX03231-23 MFLX03231-27 MFLX03231-29 MFLX03231-01 MFLX03231-03 MFLX03231-07 MFLX03231-21 MFLX03231-33 MFLX03231-41 MFLX03231-43 MFLX03231-45 MFLX03231-47 MFLX03231-51 MFLX03231-61 MFLX03231-67 MFLX03231-09
Hedland Spring-Loaded In-Line Flowmeters
Flow Meters
Provide accurate readings while mounted in any position.
  • Rugged design resists shock and vibration
  • Control pumps or valves with an optional limit switch
These economical flowmeters are built with a spring-retained piston for direct, accurate readings regardless of the position you mount them. Use them to monitor hydraulic systems or chemical processes; the meters also work well to check pump and control valve performance.
Note: Polysulfone is not suited for direct contact with sunlight.
Considering limit switches? Use to provide an output signal at aMore Product Information