Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
G-TH-075-V-3 G-TH-075-E3 G-TH-150-E-1 G-TH-150-S2 G-TH-100-V-1 G-TH-100-S-1 G-TH-200-E-1 G-TH-100-S2 G-TH-150-V-4 G-TH-200-S3 G-TH-050-S-1 G-TH-075-S3 G-TH-150-E2 G-TH-075-S-1 G-TH-200-S4 G-TH-150-V-2 G-TH-150-V-1 G-TH-075-V-1 G-TH-150-S3 G-TH-050-V-1 G-TH-075-E-1 G-TH-150-S4 G-TH-200-V-1 G-TH-100-E-1 G-TH-200-S2 G-TH-050-E-1 G-TH-075-E2 G-TH-075-S2 G-TH-150-E4 G-TH-050-S2 G-TH-200-V-2 G-TH-200-S-1 G-TH-150-V-3 G-TH-150-S-1 G-TH-050-S3
MFLX30544-12EA 221.2 CAD
MFLX30544-12 MFLX30544-14 MFLX30544-16 MFLX30542-14 MFLX30543-04 MFLX30542-13 MFLX30542-12 MFLX30542-10 MFLX30543-00 MFLX30542-09 MFLX30542-08 MFLX30542-06 MFLX30543-18 MFLX30542-05 MFLX30542-04 MFLX30543-16 MFLX30544-06 MFLX30545-12 MFLX30544-00 MFLX30545-13 MFLX30545-14 MFLX30544-04 MFLX30545-16 MFLX30544-05 MFLX30543-14 MFLX30542-02 MFLX30543-13 MFLX30542-01 MFLX30543-12 MFLX30542-00 MFLX30543-08 MFLX30542-18 MFLX30542-17 MFLX30542-16 MFLX30543-05
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Seals Gaskets
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.