Hand-Operated Rotary Drum Pumps
Supplier: Action Pump
3003T 3004 219 3005 9005H/T
MFLX06510-00EA 255.96 CAD
MFLX06510-00 MFLX06510-05 MFLX06510-10 MFLX06510-17 MFLX06510-19
Hand-Operated Rotary Drum Pumps
Pumps Drum/Transfer Pumps
Dispense a variety of liquids compatible with polypropylene, Ryton® PPS, cast iron, aluminum, stainless steel, or PTFE.
  • Lightweight and easy to use
  • Smooth, non-pulsating flow
  • Operate either in forward or reverse direction with a discharge hose
Polypropylene rotary pump (06510-00) is excellent for most organic acids, waxes, plating solutions, detergents and water soluble inorganic chemicals. Ryton PPS rotary pump (06510-05) is able to dispense aggressive chemicals such as esters, ketones and hydrocarbons. Ryton PPS pumps include a stainless steelMore Product Information