Hand-Operated Siphon Drum Pumps
Supplier: Action Pump
4005(LSS-1345) 4007 4006 5000
MFLX06422-20EA 40.45 CAD
MFLX06422-20 MFLX06439-00 MFLX06439-10 MFLX07079-70 MFLX07079-75
Hand-Operated Siphon Drum Pumps
Pumps Drum/Transfer Pumps
Economical siphon pumps are ideal for a variety of fluids.
  • Adaptable to most pails and drums
Siphon pumps are inexpensive, disposable hand pumps. Model 06439-00 features a squeeze-bulb action while models 06422-20 and 06439-10 feature a bellows action for quick and easy emptying of containers. Model 07079-70 creates a siphon with a few strokes of the pump.