Electrically and Pneumatically Actuated Elliptical Valves
Supplier: EVSCO INC
523-PP-1-RE1 223-PP-1-RA4 MFLX98153-03 MFLX98153-15 MFLX98153-30 MFLX98153-33 MFLX98154-16 MFLX98154-31 MFLX98155-14
MFLX98153-16EA 790 CAD
MFLX98153-16 MFLX98154-13 MFLX98153-03 MFLX98153-15 MFLX98153-30 MFLX98153-33 MFLX98154-16 MFLX98154-31 MFLX98155-14
Electrically and Pneumatically Actuated Elliptical Valves
Valves Elliptical Valves
Completely disassemble in-line for easy cleaning.
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73 °F (10 bar at 22 °C) (liquids).