Flowline Switch-Tek Ultrasonic Level Switches
Supplier: FLOWLINE
LU10-1305 LU10-2305 MFLX23002-15 MFLX43299-02 MFLX43299-06 MFLX43299-08
MFLX23002-14EA 897.44 CAD
MFLX23002-14 MFLX23002-16 MFLX23002-15 MFLX43299-02 MFLX43299-06 MFLX43299-08
Flowline Switch-Tek Ultrasonic Level Switches
Level Sensors
Compatible with Smart Trak and Switch-Pak installation fittings.
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.