Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Supplier: BECKSON INDUSTRIAL PRODUCTS
PC0960 CP1248S CP3648S
MFLX70607-00EA 89.59 CAD
MFLX70607-00 MFLX70607-10 MFLX70607-12
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Pumps Drum/Transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Portable and self-priming
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.