LMI Solenoid-Diaphragm Metering Remote Control Pumps
Supplier: EQUIP SOLUTIONS
B721-398SI W2T466694 C741-36 MFLX74514-45 MFLX74514-30
MFLX74514-20EA 2591.2 CAD
MFLX74514-20 MFLX74514-40 MFLX74514-50 MFLX74514-45 MFLX74514-30
LMI Solenoid-Diaphragm Metering Remote Control Pumps
Pumps Diaphragm Pumps
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
An external pulse trigger can be accepted for remote stroke frequency control. Low-flow pump models (1 GPH and lower) include easy access to 4 pin connections.
Service kits include a replacement diaphragm, seal, four pump head screws, and check valves.
Delivery information: Includes four-function valve, 16-ftMore Product Information