LMI Manually Adjustable Control Metering Pumps
Supplier: EQUIP SOLUTIONS
B121-398SI W2T464932 C131-318SI MFLX74513-25 MFLX74513-30
MFLX74513-20EA 2480.6 CAD
MFLX74513-20 MFLX74513-40 MFLX74513-25 MFLX74513-30
LMI Manually Adjustable Control Metering Pumps
Pumps Diaphragm Pumps
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
These pumps include a four-function valve — an anti-siphon, back pressure, priming, and pressure relief valve all in one.
What's included: a four-function valve, 16-ft polyethylene tubing, foot valve/strainer, and 8-ft power cord with molded plug.
Service kits include a replacement diaphragm, seal, four pump headMore Product Information