ITT Jabsco Positive Displacement Pump Pulse Dampeners
Supplier: ITT Jabsco
853327,AD-10PS 853348;AD-25PS AD-10TT (3/8" NPT)
MFLX74021-00EA 5864.96 CAD
MFLX74021-00 MFLX74021-14 MFLX74021-04
ITT Jabsco Positive Displacement Pump Pulse Dampeners
Pumps Air-Operated Pumps
Create a pulse-free flow out of your positive displacement pump.
  • Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.
More Info: These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.