High-Flow Polyester Felt-Filter Bags
Supplier: Pentair
KE10K4S KE100K3S KE25K4S
MFLX01524-84EA 9.8 CAD
MFLX01524-84 MFLX01524-82 MFLX01524-48
High-Flow Polyester Felt-Filter Bags
Filters Bag Filter Systems
Wide range of options for temperature, flow rate, and filtration quality.
  • Constructed of Polyester Felt
Designed for higher flow with an average efficiency at the rated pore size. Polyester felt is ideal for general-purpose filtration. The bags are glazed to minimize fiber migration. Choose polyester for higher temperature applications, rated for a temperature range of 300 °F (149 °C) maximum.