Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
WMD-30LFY-115 D-30FY(T)-220V MFLX72011-10 MFLX72011-25
MFLX72011-00EA 1643.2 CAD
MFLX72011-00 MFLX72011-05 MFLX72011-10 MFLX72011-25
Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
Fluoroplastic pumps with carbon fiber-reinforced ETFE are designed for highly corrosive or high-purity applications.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.