CPC® Metal Quick-Disconnect Fittings, Threaded Bodies
Supplier: VWR International
MFLX31403-00 MFLX31403-01 MFLX31403-05 MFLX31404-00 MFLX31404-20 MFLX31404-21 LCD10004 (10PK) LCD10006 (10PK) LC10004 (10PK) MC1002 (10PK) MCD1002 (10PK) MCD1004 (10PK)
MFLX31403-00EA 51.82 CAD
MFLX31403-00 MFLX31403-01 MFLX31403-05 MFLX31403-06 MFLX31404-00 MFLX31404-01 MFLX31404-20 MFLX31404-21 MFLX31403-00BLK MFLX31403-01BLK MFLX31403-05BLK MFLX31404-00BLK MFLX31404-20BLK MFLX31404-21BLK
CPC® Metal Quick-Disconnect Fittings, Threaded Bodies
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A complete coupler requires a fitting body and an insert with the same flow size (order separately).