CPC® Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: VWR International
MFLX06360-92 MFLX06360-93 MFLX31311-61 MFLX31311-62 MFLX31311-63 MFLX31311-71 MFLX31311-72 MFLX31311-73 APC22004 (25PK) APC22006 (250PK) MPC22004T03M (250PK) MPC22006T03M MPX22803M (250PK) MPC22004T39M (250PK) MPC22006T39M (250PK) MPX22839M (250PK) MPC22002T39MB25 25PK
MFLX06360-92EA 33.5 CAD
MFLX06360-92 MFLX06360-93 MFLX31311-52 MFLX61027-97 MFLX31311-61 MFLX31311-62 MFLX31311-63 MFLX31311-71 MFLX31311-72 MFLX31311-73 MFLX06360-92BLK MFLX06360-93BLK MFLX31311-61BLK MFLX31311-62BLK MFLX31311-63BLK MFLX31311-71BLK MFLX31311-72BLK MFLX31311-73BLK MFLX61027-97BLK
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Fittings and Connectors
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).