CPC® Miniature Quick-Disconnect Fittings, Hose Barb Complete Couplings
Supplier: VWR International
MFLX06363-88 MFLX06363-91 MFLX06363-93 MFLX06363-87 MFLX06363-89 MFLX06363-96 SMC01 (50PK) SMC02 (50PK) SMC0212 (50PK) SMC0112 SMCD02 50PK
MFLX06363-88EA 155.47 CAD
MFLX06363-88 MFLX06363-91 MFLX06363-92 MFLX06363-93 MFLX06363-87 MFLX06363-89 MFLX06363-96 MFLX06363-88BLK MFLX06363-91BLK MFLX06363-93BLK MFLX06363-89BLK MFLX06363-92BLK
CPC® Miniature Quick-Disconnect Fittings, Hose Barb Complete Couplings
Fittings and Connectors
No more kinked tubing—allows tubing to rotate freely.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected - preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
The complete couplings come with one insert and one body fitting.