Liquid-Solids Separator / Concentrator Units
Supplier: VWR International
MFLX29995-02 MFLX29995-06 MFLX29995-12 MFLX29995-18 MFLX29995-04 MFLX29995-08 MFLX29995-10 MFLX29995-14
MFLX29995-02EA 1627.4 CAD
MFLX29995-02 MFLX29995-06 MFLX29995-12 MFLX29995-18 MFLX29995-04 MFLX29995-08 MFLX29995-10 MFLX29995-14
Liquid-Solids Separator / Concentrator Units
Pumps Centrifugal Separators
Proven design utilizes centrifugal action to remove and concentrate solids.
  • No screens, cartridges or filters to clean or replace
  • Backflushing is not required - minimizing the loss of water and process fluids
  • Unit has no moving parts - less maintenance down time required
In many processes, removal of solids can extend the usefulness of the process media and protect equipment from fouling and abrasive wear; regulatory benefits also exist in many applications. The basic design requires no electrical power and minimal piping, which makes cost payback on the installation quick.
The unit has a low and consistent pressure loss making process integration easy. Made from carbon steel, it is extremely durable and relatively inexpensive compared to other separation technologies.