Flowline Nonintrusive RF Capacitance Level Switch
Supplier: Flowline
LP50-6005 LP50-1005
MFLX43299-05EA 415.54 CAD
MFLX43299-05 MFLX43299-12
Flowline Nonintrusive RF Capacitance Level Switch
Level Sensors
Measure level without touching fluid to avoid contamination.
  • Ideal for applications where purity is important
If you are concerned about your liquid's purity, then this nonintrusive level switch is the solution. Sensor mounts on the outside of your nonmetallic tank and never touches your liquid. Use with clean, noncoating, nonscaling, and conductive liquids. NEMA 4X polysulfone sensor enclosure is ideal for industrial environments. Includes 8-ft cable. Weld mounting bracket (sold separately) in place for firm hold.