Flowline Plastic Mini-Float Level Switches
Supplier: FLOWLINE
LV20-1201 LV20-5201
MFLX43299-24EA 88.32 CAD
MFLX43299-24 MFLX43299-26
Flowline Plastic Mini-Float Level Switches
Level Sensors
Control liquid level even in corrosive environments.
  • Excellent for clean, non-coating or non-scaling liquids
Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.
Delivery information: Includes submersible float with 2-ft (0.6-m) cable.