Flowline Switch-Pro Remote Relay Level Controller
Supplier: FLOWLINE
LC41-1001 LC42-1001 MFLX43300-30
MFLX43300-32EA 774.2 CAD
MFLX43300-32 MFLX43300-34 MFLX43300-30
Flowline Switch-Pro Remote Relay Level Controller
Level Sensors
Control large loads—SPDT relay(s) rated 250 VAC, 10 A resistive.
  • LED indicators provide user with power, sensor and relay status
  • Fail-safe relay control of pumps and valves with 0-60 second time delay
  • PP 35 mm DIN enclosure with removable terminal straps
Controllers accept all Flowline(R) point level sensors and allow you to control external devices like pumps, valves, or alarms. Adjustable time delay prevents frequent relay-cycling. LEDs indicate wet/dry sensor condition. Mount controllers on any 35-mm DIN rail.