Polypropylene and PVC Pressure Regulators
Supplier: VWR International
PRHM050V-PP PRHM050V-PV
MFLX01350-82EA 1216.6 CAD
MFLX01350-82 MFLX01350-86
Polypropylene and PVC Pressure Regulators
Valves Pressure-Relief Valves
Easily control your outlet pressure.
  • Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.