Wide-Range Pressure Relief Valves
Supplier: VWR International
MFLX01350-60 MFLX01350-62 MFLX01350-68 MFLX01350-64 MFLX01350-66 MFLX01350-70 MFLX01354-00
MFLX01350-60EA 489.8 CAD
MFLX01350-60 MFLX01350-62 MFLX01350-68 MFLX01350-64 MFLX01350-66 MFLX01350-70 MFLX01354-00
Wide-Range Pressure Relief Valves
Valves Pressure-Relief Valves
Prevent leaks with the emergency shutdown design.
  • Three-seal system vents liquid to warn of approaching maintenance
Valves with connections of 1/4" or 1/2" have a diaphragm made of Viton® and withstand a maximum inlet pressure of 210 psig. All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140 psig. The shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance. All valves are recommended for liquid applications.