+GF+ Piping Systems Metering Ball Valves
Supplier: BURT PROCESS EQUIPMENT
161523521 161523522 167523412
MFLX01360-40EA 243.32 CAD
MFLX01360-40 MFLX01360-41 MFLX01360-45
+GF+ Piping Systems Metering Ball Valves
Valves Metering Valves
Precise control over 180° range.
  • Choose between PVC or polypropylene (PP) construction
Ball valves give you precise control over the entire 180° range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5° increments.