Stopcocks with Flow Indicator, PVC
Supplier: ASAHI/AMERICA INC
#1077002 #1078002 #1079002 MFLX06225-50 MFLX06225-58
MFLX06225-52EA 65.41 CAD
MFLX06225-52 MFLX06225-54 MFLX06225-56 MFLX06225-50 MFLX06225-58
Stopcocks with Flow Indicator, PVC
Stopcocks
Easily control flow with flow indicator handle.
  • Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.