Double-Walled Heat-Shrink Tubing, PTFE
Supplier: VWR International
MFLX95810-13 MFLX95810-15 MFLX95810-16 MFLX95810-10 MFLX95810-11 MFLX95810-14
MFLX95810-13EA 711 CAD
MFLX95810-13 MFLX95810-15 MFLX95810-16 MFLX95810-10 MFLX95810-11 MFLX95810-14
Double-Walled Heat-Shrink Tubing, PTFE
Tubing Heat-Shrink Tubing
Ideal for chemically inert applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.