Standard-Wall PTFE Heat-Shrink Transfer Tubing
Supplier: VWR International
MFLX06851-22 MFLX06851-42 MFLX06851-12 MFLX06851-32 MFLX06851-52
MFLX06851-22EA 144.41 CAD
MFLX06851-22 MFLX06851-42 MFLX06851-12 MFLX06851-32 MFLX06851-52
Standard-Wall PTFE Heat-Shrink Transfer Tubing
Tubing Heat-Shrink Tubing
Ideal for chemically inert applications.
  • High electrical and chemical resistance
  • Comes in 3-ft lengths
  • Opaque
Shrink ratio: 2:1
Recovery temperature: 621 °F (327 °C)
Characteristics: Opaque; high electrical and chemical resistance.