Direct Coupled 303 Stainless Steel Rotary Vane Pumps
Supplier: BERNS CORP
PA411-50MT PA811-75MT
MFLX79411-15EA 2227.8 CAD
MFLX79411-15 MFLX79411-10
Direct Coupled 303 Stainless Steel Rotary Vane Pumps
Pumps Flexible/Rotary/Roller Impeller Pumps
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.