Direct Coupled Brass-Bodied Rotary Vane Pumps
Supplier: BERNS CORP
PO301X-33M PA401X-50M PA801-75MT MFLX79410-00
MFLX79410-05EA 837.4 CAD
MFLX79410-05 MFLX79410-10 MFLX79410-15 MFLX79410-00
Direct Coupled Brass-Bodied Rotary Vane Pumps
Pumps Flexible/Rotary/Roller Impeller Pumps
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.