Nordel Flexible Liner Pumps, Polypropylene
Supplier: VANTON PUMP & EQUIPMENT CORP
CC-PY6B/E, TEFC CC-PY60B/E, TEFC MFLX79440-10 MFLX79440-20 MFLX79441-10 MFLX79441-20
MFLX79440-00EA 1706.4 CAD
MFLX79440-00 MFLX79440-30 MFLX79440-10 MFLX79440-20 MFLX79441-10 MFLX79441-20
Nordel Flexible Liner Pumps, Polypropylene
Pumps Diaphragm Pumps
Unique sealless design permits gentle pumping action that provides little churning or foaming.
  • Transfer liquids containing up to 20% soft solids
  • Sealless design with secondary containment system prevents potentially hazardous leakage
  • Fixed-speed pumps are self-priming, reversible, and can run dry without damage
Polypropylene pumps have a Nordel® rubber liner; PTFE pumps have a Hypalon® rubber liner. Variable-speed models and DC-powered pumps are available — call our Applications Specialists for more information.