Plastic Hand-Operated Siphon/Lift Drum Pump
Supplier: BECKSON INDUSTRIAL PRODUCTS
MP1848S
MFLX06437-10EA 140.94 CAD
MFLX06437-10
Plastic Hand-Operated Siphon/Lift Drum Pump
Pumps Drum/Transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Equipped with a 2" IPS bung adapter for pumping detergents, water, dilute alkalies, or waxes from drums
  • Cut inlet tube to fit your container
This plastic hand-operated drum pump is ideal for pumping detergents, water, dilute alkalies, or waxes from drums. Constructed with a PVC body and hose, Hypalon valves and seals, and an LDPE tube. This pump offers 16 strokes/gallon (5 strokes/liter).