Plastic Hand-Operated Siphon/Lift Pail Pump
Supplier: BECKSON INDUSTRIAL PRODUCTS
9A
MFLX06437-00EA 115.81 CAD
MFLX06437-00
Plastic Hand-Operated Siphon/Lift Pail Pump
Pumps Drum/Transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Suitable for pumping petroleum products or water
  • Cut inlet tube to fit your container
  • Maximum suction lift is 10 ft (3.0-m)
This hand-operated drum pump is ideal for pumping petroleum products or water. Constructed of PVC barrel and hose, urethane-buna valves and seals, this pump offers 32 strokes/gallon (9 strokes/liter).