Hand-Operated Piston Dispensing Drum Pump, PVC
Supplier: Action Pump
505 504 506 MFLX07079-65
MFLX07079-50EA 111.07 CAD
MFLX07079-50 MFLX07079-55 MFLX07079-60 MFLX07079-65
Hand-Operated Piston Dispensing Drum Pump, PVC
Pumps Drum/Transfer Pumps
Economical piston pumps are ideal for a variety of fluids.
  • Adjustable to most pails and drums
Models 07079-50 and -55 feature an angled spout to help prevent dripping. Models 07079-60 and -65 are made entirely of PVC, including the piston and seal, making them an ideal option for sanitary applications.