Micropump® A-Mount Suction Shoe Gear Pump Heads
Supplier: MICROPUMP INC
81726 81112 81113 81114 81282 L16703 L20561 L3468 L20562 MI0022 / ZP-200 / 81281 MI0023 / ZP-201 / 81282 MI0306 / Z-200P / 220004 220001 MFLX07003-02
MFLX07002-14EA 2212 CAD
MFLX07002-14 MFLX07002-25 MFLX07002-26 MFLX07002-27 MFLX07003-04 MFLX73003-14 MFLX73004-00 MFLX73004-02 MFLX73004-04 MFLX74004-19 MFLX74004-21 MFLX74004-77 MFLX74012-51 MFLX07003-02
Micropump® A-Mount Suction Shoe Gear Pump Heads
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.