Magnetic Drive Centrifugal Pumps with Open Drip Proof (ODP) Motor
Supplier: MARCH MANUFACTURING INC
0135-0163-0410 0125-0069-0100 0921-0007-0100 0125-0069-0300 0921-0007-0200 0921-0007-0300 0130-0018-0210 0150-0136-0110 MFLX07022-20 MFLX07022-23 MFLX07023-05 MFLX07023-06 MFLX07023-13
MFLX07004-32EA 511.92 CAD
MFLX07004-32 MFLX07021-00 MFLX07021-01 MFLX07021-02 MFLX07021-03 MFLX07022-21 MFLX07022-22 MFLX07023-12 MFLX07022-20 MFLX07022-23 MFLX07023-05 MFLX07023-06 MFLX07023-13
Magnetic Drive Centrifugal Pumps with Open Drip Proof (ODP) Motor
Pumps Centrifugal Pumps
Polypropylene (PP)-encapsulated magnets offer excellent chemical resistance.
  • Easily disassemble pumps for cleaning
  • ODP pumps provide an economical option for use in chemical OEM and industrial applications
These magnetic drive centrifugal pumps with open drip proof (ODP) motors are excellent for applications that require chemical resistance. Pumps are constructed with a polypropylene (PP) body and impeller, Viton® O-ring, and PP-encapsulated ceramic spindle. Maximum temperature of 190˚F (88˚C).
At 60 Hz; flow rates and ft of head are 5/6 these values at 50 Hz.
Delivery information: Pumps include mounting base and 3-ft (1 m), three-wire cord (no plug). Service kits (sold separately under "Accessories") include impeller/magnet assembly and VITON® O-ring.