Mini Submersible Magnetic Drive Pumps
Supplier: MARCH MANUFACTURING INC
0115-0007-0200 0125-0057-0200 130-158-02 0115-0007-0800 MFLX07105-07 MFLX07142-05 MFLX07142-16 MFLX07142-17 MFLX07142-31
MFLX07105-05EA 481.9 CAD
MFLX07105-05 MFLX07142-02 MFLX07142-15 MFLX07142-23 MFLX07105-07 MFLX07142-05 MFLX07142-16 MFLX07142-17 MFLX07142-31
Mini Submersible Magnetic Drive Pumps
Pumps Centrifugal Pumps
Operate fully submerged or in open air.
  • Liquid-cooled models keep heat-sensitive fluids from warming up
  • Thick (1/8") epoxy coating dissipates heat quickly and provides good chemical resistance
  • UL-Listed
These mini submersible magnetic drive pumps are good for low-viscosity liquids including solvents, dyes, inks, and mild corrosives.
Delivery information: Includes a three-wire cord.