Gems LS-3 Series Level Switches
Supplier: VWR International
MFLX07187-02 MFLX07187-04 MFLX07187-06 MFLX07187-08 MFLX07187-10 MFLX07187-14 MFLX07187-18 MFLX07187-20 MFLX07188-40 MFLX07188-35 MFLX07187-12 MFLX07187-16
MFLX07187-02EA 131.77 CAD
MFLX07187-02 MFLX07187-04 MFLX07187-06 MFLX07187-08 MFLX07187-10 MFLX07187-14 MFLX07187-18 MFLX07187-20 MFLX07188-40 MFLX07188-35 MFLX07187-12 MFLX07187-16
Gems LS-3 Series Level Switches
Level Sensors
Install from the top of your tank—attach your own conduit to protect wiring.
  • Get longer life and more durability with hermetically sealed switches
All models accept 120/240 VAC and operate as NO or NC switches except where noted. Switches include 24" leads (except 07187-12 which has 72" leads). If your level control system exceeds the current rating in the table, use a controller with your switch.