GF Signet Installation Tee Fittings, CPVC/PVC 80
Supplier: BURT PROCESS EQUIPMENT
MPV8T005F MPV8T010F MVP8T015F MPV8T020 PV8T040 MCPV8T010F MFLX05620-22 MFLX05620-24 MFLX05620-27 MFLX05620-28 MFLX05620-31 MFLX05620-34 MFLX05620-35
MFLX05620-21EA 276.5 CAD
MFLX05620-21 MFLX05620-23 MFLX05620-25 MFLX05620-26 MFLX05620-29 MFLX05620-33 MFLX05620-22 MFLX05620-24 MFLX05620-27 MFLX05620-28 MFLX05620-31 MFLX05620-34 MFLX05620-35
GF Signet Installation Tee Fittings, CPVC/PVC 80
Flow Meters
Easily install your flowmeters.
  • Constructed of durable CPVC/PVC 80
PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.