Rotor-X Low-Flow Paddle-Wheel Flow Sensors
Supplier: BURT PROCESS EQUIPMENT
3-2536-P0 3-2536-P1 3-2536-V0
MFLX32500-00EA 1398.3 CAD
MFLX32500-00 MFLX32500-02 MFLX32500-10
Rotor-X Low-Flow Paddle-Wheel Flow Sensors
Flow Meters
Measure low flow rates down to 0.3 ft/second.
  • Eliminate cavitation and minimize pressure drop
Measure low flow rates in almost any size pipe. These sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a Hall effect sensor, which generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 1000 feet without amplification.
The open-core design eliminates cavitation at flow velocities from 0.3 to 20 ft/second. Closed or solid paddle-wheel designsMore Product Information