ReSeal Stainless Steel Sanitary Couplings
Supplier: VWR International
MFLX31511-07 MFLX31511-13 MFLX31511-20
MFLX31511-07EA 722.06 CAD
MFLX31511-07 MFLX31511-13 MFLX31511-20
ReSeal Stainless Steel Sanitary Couplings
Fittings and Connectors
Meets USDA compliance and 3A Sanitary Standards, and is FDA compliant.
  • Constructed of durable stainless steel with PTFE O-ring
Unlike permanent couplers, these couplers can be attached to a new length of tubing when the old tubing wears out. Suitable for clean-in-place, steam-in-place, and autoclaving for reuse; compatible with TRI-CLAMP® fittings. The internal stem is made of 316L stainless steel with an interior surface finish of 25RA or better. Fittings also feature an external sleeve made from a high-temperature, high-strength polymer and a PTFE O-ring to protect against any external entrapment. Compression seal eliminates wicking between couplers and tubing.
Use with flexible. Step-up couplers are available upon request—call our Application Specialists for details.
Listed silicone tubing is peroxide cured-see "Tubing" on pages 1140-1155 to order platinum-cured silicone tubing.